4-Nitro-m-cresol
Catalog # APOSOR8262-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:127…129 °C
:153.14 g/mol
Storage Temperature:Ambient
CAS Number:2581-34-2
Formula:C₇H₇NO₃
MDL Number:MFCD00007333