
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine 98%
Catalog # APOSOR8250-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.6 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:929000-42-0
Formula:C₇H₆ClN₃
MDL Number:MFCD09258771