
6-Nitro-m-cresol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:700-38-9
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007111
:153.14 g/mol
Melting Pt:53…56 °C
Flash Pt:109 °C
Storage Temperature:Ambient
EINECS:211-843-0
UN:2446
ADR:6.1,III
:GHS07
Frequently Bought Together