
3-(2-Methylpyrimidin-4-yl)aniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175201-90-8
:
Formula:C₁₁H₁₁N₃
MDL Number:MFCD00067950
:185.23 g/mol
Melting Pt:107…110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together