2,4-Di-tert-butoxy-5-pyrimidineboronic acid hydrate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306935-93-3
:
Formula:C₁₂H₂₁BN₂O₄·H₂O
MDL Number:MFCD02180675
Melting Pt:250 °C
Storage Temperature:Refrigerator
Frequently Bought Together