
1-Phenyl-2-pyrazin-2-ylethanone 95+%
Catalog # APOSOR7947-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.23 g/mol
Boiling Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
CAS Number:40061-45-8
Formula:C₁₂H₁₀N₂O
MDL Number:MFCD04037906