Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
Irritant
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
Catalog # APOSOR7929-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
Catalog # APOSOR7929-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.29 g/mol
Melting Pt:163…166 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:16223-25-9
Formula:C₁₃H₁₆N₂O
MDL Number:MFCD03426085