
4-(4-Nitrophenyl)-1H-pyrazole 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114474-26-9
:
Formula:C₉H₇N₃O₂
MDL Number:MFCD04037972
:189.18 g/mol
Melting Pt:205…208 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together