
Indolin-1-amine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:92259-86-4
:
Formula:C₈H₁₁ClN₂
MDL Number:MFCD04037889
:170.64 g/mol
Melting Pt:185 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together