
4-Bromomethyl-1,2-dihydroquinoline-2-one 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4876-10-2
:
Formula:C₁₀H₈BrNO
MDL Number:MFCD03426152
:238.08 g/mol
Melting Pt:252 °C (dec.)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together