2-Methoxy-4,6-dimethylnicotinonitrile
Catalog # APOSOR7502-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.19 g/mol
Storage Temperature:Ambient
CAS Number:65515-39-1
Formula:C₉H₁₀N₂O
MDL Number:MFCD00124716