Order Entry
Czech Republic
ContactUsLinkComponent
4'-(3-Methoxyphenyl)acetophenone
Irritant
4'-(3-Methoxyphenyl)acetophenone
Catalog # APOSOR7476-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-(3-Methoxyphenyl)acetophenone
Catalog # APOSOR7476-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:76650-30-1
Formula:C₁₅H₁₄O₂
MDL Number:MFCD04039105