
4'-Formylbiphenyl-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:70916-98-2
:
Formula:C₁₄H₁₀O₃
MDL Number:MFCD03424613
:226.2274 g/mol
Melting Pt:150 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together