Order Entry
Czech Republic
ContactUsLinkComponent
4'-Chlorobiphenyl-4-carboxaldehyde 97%
Irritant
4'-Chlorobiphenyl-4-carboxaldehyde 97%
Catalog # APOSOR7357-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Chlorobiphenyl-4-carboxaldehyde 97%
Catalog # APOSOR7357-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :216.67 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:80565-30-6
Formula:C₁₃H₉ClO
MDL Number:MFCD01631911