
2-Methoxy-5-pyridineboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:163105-89-3
:
Formula:C₆H₈BNO₃
MDL Number:MFCD02093044
:152.95 g/mol
Melting Pt:135…140 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together