
Dicyclopropylamine hydrochloride
Catalog # APOSOR7058-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:133.62 g/mol
Melting Pt:92…98 °C
:GHS07
Storage Temperature:Ambient
CAS Number:246257-69-2
Formula:C₆H₁₂ClN
MDL Number:MFCD06796385