
4-Bromo-N,N-diethyl-3-methoxybenzamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:889676-36-2
:
Formula:C₁₂H₁₆BrNO₂
MDL Number:MFCD08436037
Melting Pt:68…70 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together