

1,3-Bis(diphenylphosphino)propanenickel(II) chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15629-92-2
:
Formula:C₂₇H₂₆Cl₂NiP₂
MDL Number:MFCD00015318
:542.042 g/mol
Melting Pt:213 °C (dec.)
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together