
2-(Di-tert-butylphosphino)biphenyl 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:224311-51-7
:
Formula:C₂₀H₂₇P
MDL Number:MFCD01862440
:298.40 g/mol
Melting Pt:84…88 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together