
Methyl-3-formyl-4-hydroxybenzoate
Catalog # APOSOR6967-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:180.16 g/mol
Melting Pt:80…84 °C
:GHS07
Storage Temperature:Ambient
CAS Number:24589-99-9
Formula:C₉H₈O₄
MDL Number:MFCD01661473