

1-(2-Phenylethyl)piperazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5321-49-3
:
Formula:C₁₂H₁₈N₂
MDL Number:MFCD00040740
:190.29 g/mol
Boiling Pt:276 °C
Density:1.006 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:226-186-5
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Frequently Bought Together