
2-(1,3-Oxazol-5-yl)aniline 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:774238-36-7
:
Formula:C₉H₈N₂O
MDL Number:MFCD07772822
Melting Pt:53…56 °C
UN:2811
ADR:6.1,III
:160.172 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together