
6-Methoxy-1-tetralone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1078-19-9
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001695
Boiling Pt:171 °C/11mmHg
Melting Pt:76…79 °C
:176.22 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together