Ethyl-1-piperidinepropionate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(Piperidino)propionic Acid Ethyl Ester, Ethyl 3-(Piperidino)propionate, 1-Piperidinepropionic Acid Ethyl Ester
CAS Number:19653-33-9
:
Formula:C₁₀H₁₉NO₂
MDL Number:MFCD00006514
Boiling Pt:108…110 °C
Molecular Weight:185.27 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together