
tert-Butyl-3-bromobenzoate 95+%
Catalog # APOSOR6394-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:81…83 °C/0.2mmHg
:257.1200 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:69038-74-0
Formula:C₁₁H₁₃BrO₂
MDL Number:MFCD06657802