
2-Aminopyrimidine-5-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3167-50-8
:
Formula:C₅H₅N₃O₂
MDL Number:MFCD05864791
:139.1121 g/mol
Melting Pt:335…340 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together