
4-(1H-Pyrazol-4-yl)benzoic acid 95+%
Catalog # APOSOR6230-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:188.19 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1017794-47-6
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD06803014