
Thiomorpholine-1,1-dioxide hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59801-62-6
:
Formula:C₄H₁₀ClNO₂S
MDL Number:MFCD06738995
:171.6457 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together