Order Entry
Czech Republic
ContactUsLinkComponent
2-Aminomalonamide 97%
2-Aminomalonamide 97%
Catalog # APOSOR61428-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Aminomalonamide 97%
Catalog # APOSOR61428-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :117.1100 g/mol
Melting Pt:192…196 °C
Storage Temperature:Ambient
CAS Number:62009-47-6
Formula:C₃H₇N₃O₂
MDL Number:MFCD00015932