
ω,3,4-Trichloroacetophenone 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42981-08-8
:
Formula:C₈H₅Cl₃O
MDL Number:MFCD03970382
Boiling Pt:171…173 °C/15mmHg
Melting Pt:46…47 °C
UN:1759
ADR:8,III
:223.49 g/mol
:GHS05
Storage Temperature:Ambient
Frequently Bought Together