Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxypyrimidine hydrochloride
2-Hydroxypyrimidine hydrochloride
2-Hydroxypyrimidine hydrochloride
APOSOR61010-1G
:
Apollo Scientific
:
2-Hydroxypyrimidine hydrochloride
APOSOR61010-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
132.55 g/mol
:
200…205 °C (dec.)
:
253-897-8
:
GHS07
:
Ambient
:
38353-09-2
:
C₄H₄N₂O
:
MFCD00012781