
1-[(Ammoniooxy)methyl]-3-chlorobenzene chloride
Catalog # APOSOR6062-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:206…212 °C
:GHS07
Storage Temperature:Ambient
CAS Number:29605-78-5
Formula:C₇H₉Cl₂NO
MDL Number:MFCD01114569