1-[(Ammoniooxy)methyl]-2-bromobenzene chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51572-91-9
:
Formula:C₇H₉BrClNO
MDL Number:MFCD01114573
Melting Pt:144…146 °C
Storage Temperature:Ambient
Frequently Bought Together