Order Entry
Czech Republic
ContactUsLinkComponent
2-Aminopyrimidine-5-carbonitrile 97%
Irritant
2-Aminopyrimidine-5-carbonitrile 97%
Catalog # APOSOR60250-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Aminopyrimidine-5-carbonitrile 97%
Catalog # APOSOR60250-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :120.1121 g/mol
Melting Pt:275 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1753-48-6
Formula:C₅H₄N₄
MDL Number:MFCD00232791