3-Hydroxypropionic acid 30% in aqueous solution
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid
- CAS Number:503-66-2
- Formula:C₃H₆O₃
- MDL Number:MFCD00058998
- Molecular Weight:90.08 g/mol
- Density:1.08 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- EINECS:207-974-8
2 Options Available Below
Product Details & Documents
Synonyms: 3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid
- CAS Number:503-66-2
- Formula:C₃H₆O₃
- MDL Number:MFCD00058998
- Molecular Weight:90.08 g/mol
- Density:1.08 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- EINECS:207-974-8
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



