
2-Hydroxy-4-methylpyrimidine hydrochloride
Catalog # APOSOR59839-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:146.57 g/mol
Melting Pt:243 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5348-51-6
:MFCD00012782
Formula:C₅H₇ClN₂O