
Methyl-3-cyanobenzoate
:13531-48-1
:
:C₉H₇NO₂
:MFCD00220156
:161.1574 g/mol
:58…60 °C
:Ambient
:3439
:6.1,III
:GHS07
Frequently Bought Together
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethylacetát ≥99%, GPR RECTAPUR®
: VWR Chemicals
Cyklohexanon ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dichlormethan ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals