
3-(1H-Imidazol-1-yl)pyrazine-2-carbonitrile
Catalog # APOSOR5978-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:171.16 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:342412-44-6
Formula:C₈H₅N₅
MDL Number:MFCD09414743