
4-[4-(2-Hydroxyethyl)-1-piperazinylcarbonyl]benzeneboronic acid hydrochloride 97%
Catalog # APOSOR5961-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:314.57 g/mol
Melting Pt:100…102 °C
:GHS07
Storage Temperature:Ambient
CAS Number:913835-44-6
Formula:C₁₃H₁₉BN₂O₄·HCl
MDL Number:MFCD09027218