
2-Amino-p-cresol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:95-84-1
:
Formula:C₇H₉NO
MDL Number:MFCD00007699
:123.15 g/mol
Melting Pt:133…138 °C
EINECS:202-457-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together