
Disodium naphthalene-1,5-disulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1655-29-4
:
Formula:C₁₀H₆Na₂O₆S₂
MDL Number:MFCD00064178
:332.27 g/mol
EINECS:216-732-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together