1-(3-Aminopyridin-4-yl)ethanone
Catalog # APOSOR59461-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:136.15 g/mol
Storage Temperature:Ambient
CAS Number:13210-52-1
Formula:C₇H₈N₂O
MDL Number:MFCD07375078