N'-(4,4-Dimethylcyclohexylidene)-2,4,6-trimethylbenzenesulphonohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957066-12-5
:
Formula:C₁₇H₂₆N₂O₂S
MDL Number:MFCD09878366
Melting Pt:135…137 °C
Storage Temperature:Refrigerator
Frequently Bought Together