Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Dimethylphenylhydrazinium hydrochloride 97%
Irritant
2,3-Dimethylphenylhydrazinium hydrochloride 97%
Catalog # APOSOR5938-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dimethylphenylhydrazinium hydrochloride 97%
Catalog # APOSOR5938-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.66 g/mol
Melting Pt:210 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:56737-75-8
 :123333-92-6
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00013380