3-Chloro-N-2-dimethylaniline hydrochloride
Catalog # APOSOR59371-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:957062-82-7
Formula:C₈H₁₁Cl₂N
MDL Number:MFCD09878314