4-Methoxy-2-nitrophenylboronic acid
Catalog # APOSOR59360-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:196.950 g/mol
Storage Temperature:Refrigerator
CAS Number:860034-09-9
Formula:C₇H₈BNO₅
MDL Number:MFCD09878340