Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Hydroxy-4-(pyridin-3-yl)coumarin
7-Hydroxy-4-(pyridin-3-yl)coumarin 95+%
7-Hydroxy-4-(pyridin-3-yl)coumarin 95+%
APOSOR5886-1G
:
Apollo Scientific
:
7-Hydroxy-4-(pyridin-3-yl)coumarin 95+%
APOSOR5886-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
239.23 g/mol
:
>230 °C (dec.)
:
GHS07
:
Ambient
:
109512-76-7
:
C₁₄H₉NO₃
:
MFCD01941325
:
>230 °C (dec.)