
N,N-Diethyl-p-toluidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:613-48-9
:
Formula:C₁₁H₁₇N
MDL Number:MFCD00015113
:163.26 g/mol
Boiling Pt:230 °C
Density:0.92 g/cm³ (25 °C)
Flash Pt:102 °C
Storage Temperature:Ambient
EINECS:210-345-0
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together