
N,N-Diethyl 3-boronobenzenesulphonamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:871329-58-7
:
Formula:C₁₀H₁₆BNO₄S
MDL Number:MFCD07363746
Melting Pt:120…123 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together