1-Pyridin-4-yl-N-(pyridin-4-ylmethyl)methanamine 95+%
Catalog # APOSOR5676-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:199.26 g/mol
Storage Temperature:Ambient
CAS Number:1539-39-5
Formula:C₁₂H₁₃N₃
MDL Number:MFCD00868302